Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-6-chloro-2-fluoroiodobenzene ≥97%

Supplier: AOB CHEM USA
Warning

56273-1G 56273-25G 56273-5G
76891-876EA 62.6 USD InStock
76891-876 76891-878 76891-880
3-Bromo-6-chloro-2-fluoroiodobenzene ≥97%
3-Bromo-6-chloro-2-fluoroiodobenzene
Precaución: for Research Use Only


Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28122825
CAS Number: 1820641-80-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty