Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

57234-1G 57234-25G 57234-5G
76891-990EA 114.58 USD InStock
76891-990 76891-992 76891-994
1-Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥97% research grade
1-Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid


Formula: C₈H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03419332
CAS Number: 108444-24-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty