Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(methylsulfonyl)benzotrifluoride 95% research grade

Supplier: AOB CHEM USA
Warning

57684-1G 57684-25G 57684-5G 57684-500MG
76892-056EA 88.06 USD InStock
76892-056 76892-058 76892-060 77651-738
3-Bromo-5-(methylsulfonyl)benzotrifluoride 95% research grade
3-Bromo-5-(methylsulphonyl)benzotrifluoride


Formula: C₈H₆BrF₃O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD15143584
CAS Number: 1215205-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty