Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-Methoxy-5-(Methylsulfonyl)Benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

62552-1G 62552-25G 62552-5G 62552-500MG
76892-600EA 64.72 USD InStock
76892-600 76892-602 76892-604 77651-852
1-Bromo-3-Methoxy-5-(Methylsulfonyl)Benzene ≥97% research grade
1-Bromo-3-Methoxy-5-(Methylsulfonyl)Benzene
Precaución: for Research Use Only


Formula: C₈H₉BrO₃S
MW: 265.1242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760897
CAS Number: 2757007-98-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty