Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Aminothiazol-5-yl)-6-methylphenol ≥97%

Supplier: AOB CHEM USA
Warning

62695-1G 62695-25G 62695-500MG 62695-5G
76892-614EA 441.34 USD InStock
76892-614 76892-616 76892-618 76892-620
2-(2-Aminothiazol-5-yl)-6-methylphenol ≥97%
2-(2-Aminothiazol-5-yl)-6-methylphenol


Formula: C₁₀H₁₀N₂OS
Storage Temperature: Ambient
CAS Number: 2764728-23-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty