Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,5-Difluoro-4-methoxyphenyl)ethanone ≥95%

Supplier: AOB CHEM USA
Warning

62814-1G 62814-25G 62814-5G
76892-638EA 70.02 USD InStock
76892-638 76892-640 76892-642
1-(2,5-Difluoro-4-methoxyphenyl)ethanone ≥95%
1-(2,5-Difluoro-4-methoxyphenyl)ethanone


Formula: C₉H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD26595945
CAS Number: 1010800-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty