Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Methylphenoxy)aniline ≥97% research grade

Supplier: AOB CHEM USA
Warning

78658-1G 78658-25G 78658-5G
76893-580EA 79.57 USD InStock
76893-580 76893-582 76893-584
2-(3-Methylphenoxy)aniline ≥97% research grade
2-(3-Methylphenoxy)aniline


Formula: C₁₃H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD04610242
CAS Number: 60287-67-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty