Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Fluoro-3-Isopropyl-5-Methylphenyl)(Methyl)Sulfane ≥97% research grade

Supplier: AOB CHEM USA
Warning

85358-1G 85358-500MG 85358-5G 85358-25G
76893-740EA 2504.79 USD InStock
76893-740 76893-742 77633-780 77660-420
(4-Fluoro-3-Isopropyl-5-Methylphenyl)(Methyl)Sulfane ≥97% research grade
(4-Fluoro-3-Isopropyl-5-Methylphenyl)(Methyl)Sulfane
Precaución: for Research Use Only


Formula: C₁₁H₁₅FS
Storage Temperature: Ambient
MDL Number: MFCD34770430
CAS Number: 2918838-20-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty