Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-dimethylamino-pyrimidine-5-boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

10151-1G 10151-25G 10151-5G
76906-286EA 64.72 USD InStock
76906-286 76906-288 76906-290
4-Benzyloxy-2-dimethylamino-pyrimidine-5-boronic acid ≥97% research grade
4-Benzyloxy-2-dimethylamino-pyrimidine-5-boronic acid


Formula: C₁₃H₁₆BN₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08689616
CAS Number: 205672-21-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty