Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,5-dimethybenzeneboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

10365-1G 10365-25G 10365-5G
76906-368EA 66.84 USD InStock
76906-368 76906-370 76906-372
4-Bromo-2,5-dimethybenzeneboronic acid ≥97% research grade
4-Bromo-2,5-dimethybenzeneboronic acid


Formula: C₈H₁₀BBrO₂
MW: 228.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115647
CAS Number: 130870-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty