Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Butoxycarbonylphenylboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

11039-25G 11039-5G
76906-592EA 275.84 USD InStock
76906-592 76906-594
3-Butoxycarbonylphenylboronic acid ≥95% research grade
3-Butoxycarbonylphenylboronic acid


Formula: C₁₁H₁₅BO₄
MW: 222.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179446
CAS Number: 827300-04-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty