Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyano-5-(trifluoromethyl)benzeneboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

11256-1G 11256-25G 11256-5G
76906-688EA 102.91 USD InStock
76906-688 76906-690 76906-692
3-Cyano-5-(trifluoromethyl)benzeneboronic acid ≥95% research grade
3-Cyano-5-(trifluoromethyl)benzeneboronic acid


Formula: C₈H₅BF₃NO₂
MW: 214.938 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095102
CAS Number: 1212021-62-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty