Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Cyclopropylmethoxy)phenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

11459-1G 11459-25G 11459-500MG 11459-5G
76906-754EA 124.13 USD InStock
76906-754 76906-756 76906-758 76906-760
3-(Cyclopropylmethoxy)phenylboronic acid ≥97%
3-(Cyclopropylmethoxy)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD04039016
CAS Number: 411229-76-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty