Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methoxy-5-pyridineboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

11463-1G 11463-25G 11463-5G
76906-768EA 79.57 USD InStock
76906-768 76906-770 76906-772
3-Chloro-2-methoxy-5-pyridineboronic acid ≥97%
3-Chloro-2-methoxy-5-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₆H₇BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04972386
CAS Number: 942438-89-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty