Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dibromoisophthalic acid ≥97%

Supplier: AOB CHEM USA
Warning

119254-1G 119254-25G 119254-5G
76906-852EA 105.03 USD InStock
76906-852 76906-854 76906-856
4,6-Dibromoisophthalic acid ≥97%
4,6-Dibromoisophthalic acid


Formula: C₈H₄Br₂O₄
Storage Temperature: Ambient
MDL Number: MFCD32641193
CAS Number: 24063-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty