Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-(4-methoxybenzyl)-1H-pyrazole ≥95% research grade

Supplier: AOB CHEM USA
Warning

126399-1G 126399-25G 126399-500MG 126399-5G
76907-028EA 143.23 USD InStock
76907-028 76907-030 76907-032 76907-034
4-Bromo-1-(4-methoxybenzyl)-1H-pyrazole ≥95% research grade
4-Bromo-1-(4-methoxybenzyl)-1H-pyrazole


Formula: C₁₁H₁₁BrN₂O
MW: 267.122 g/mol
Storage Temperature: Ambient
CAS Number: 1178371-93-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty