Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetoxyphenylboronic acid, pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

12669-25G 12669-5G
76907-040EA 559.1 USD InStock
76907-040 76907-042
3-Acetoxyphenylboronic acid, pinacol ester ≥97% research grade
3-Acetoxyphenylboronic acid, pinacol ester


Formula: C₁₄H₁₉BO₄
Storage Temperature: Ambient
MDL Number: MFCD03453061
CAS Number: 480424-69-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty