Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-methylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

14397-25G 14397-5G
76907-526EA 240.83 USD InStock
76907-526 76907-528
4-Benzyloxy-2-methylbenzaldehyde ≥97%
4-Benzyloxy-2-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₅H₁₄O₂
MW: 226.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317809
CAS Number: 101093-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty