Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-methylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

14543-1G 14543-25G 14543-500MG 14543-5G
76907-608EA 185.66 USD InStock
76907-608 76907-610 76907-612 76907-614
4-Benzyloxy-2-methylbenzoic acid ≥97% research grade
4-Benzyloxy-2-methylbenzoic acid


Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00598375
CAS Number: 17819-91-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty