Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hydroxymethyl)-5-methylphenylboronic Acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

15366-1G 15366-25G 15366-500MG 15366-5G
76907-902EA 153.84 USD InStock
76907-902 76907-904 76907-906 76907-908
3-(Hydroxymethyl)-5-methylphenylboronic Acid ≥97% research grade
3-(Hydroxymethyl)-5-methylphenylboronic Acid


Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295237
CAS Number: 1451391-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty