Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-2,4,6-trifluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

15468-1G 15468-25G 15468-5G
76908-082EA 114.58 USD InStock
76908-082 76908-084 76908-086
3-Methoxy-2,4,6-trifluorophenylboronic acid ≥97%
3-Methoxy-2,4,6-trifluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD07369755
CAS Number: 849062-08-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty