Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-formyl-4-methylphenyboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

16453-1G 16453-25G 16453-500MG 16453-5G
76908-528EA 194.15 USD InStock
76908-528 76908-530 76908-532 76908-534
3-Fluoro-2-formyl-4-methylphenyboronic acid ≥97% research grade
3-Fluoro-2-formyl-4-methylphenyboronic acid


Formula: C₈H₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD13181655
CAS Number: 1451391-40-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty