Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-(Benzyloxy)-5-bromophenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

16871-1G 16871-25G 16871-500MG 16871-5G
76908-670EA 134.74 USD InStock
76908-670 76908-672 76908-674 76908-676
(3-(Benzyloxy)-5-bromophenyl)boronic acid ≥97% research grade
(3-(Benzyloxy)-5-bromophenyl)boronic acid


Formula: C₁₃H₁₂BBrO₃
MW: 306.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18087690
CAS Number: 1256355-21-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty