Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

17336-1G 17336-25G 17336-5G
76908-944EA 88.06 USD InStock
76908-944 76908-946 76908-948
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid ≥97%
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₇BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235059
CAS Number: 874219-42-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty