Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Diethylcarbamoyl-2-fluorobenzeneboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

18176-1G 18176-25G 18176-500MG 18176-5G
76909-200EA 320.4 USD InStock
76909-200 76909-202 76909-204 76909-206
3-Diethylcarbamoyl-2-fluorobenzeneboronic acid ≥97%
3-Diethylcarbamoyl-2-fluorobenzeneboronic acid


Formula: C₁₁H₁₅BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD10699633
CAS Number: 1072946-28-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty