Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

18282-1G 18282-25G 18282-5G
76909-238EA 84.88 USD InStock
76909-238 76909-240 76909-242
4-Amino-3-methoxyphenylboronic acid pinacol ester ≥97%
4-Amino-3-methoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₂₀BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06795657
CAS Number: 461699-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty