Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-(methylthio)-2-(trifluoromethyl)-benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

18503-1G 18503-25G 18503-5G
76909-338EA 64.72 USD InStock
76909-338 76909-340 76909-342
4-Bromo-1-(methylthio)-2-(trifluoromethyl)-benzene ≥97% research grade
4-Bromo-1-(methylthio)-2-(trifluoromethyl)-benzene


Formula: C₈H₆BrF₃S
Storage Temperature: Refrigerator
MDL Number: MFCD15527239
CAS Number: 300356-31-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty