Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

18756-1G 18756-25G 18756-500MG 18756-5G
76909-474EA 310.85 USD InStock
76909-474 76909-476 76909-478 76909-480
4-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid ≥97% research grade
4-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid


Formula: C₉H₈BF₃O₄
MW: 247.964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181653
CAS Number: 1451375-04-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty