Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carbamoyl-4-chlorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

18954-1G 18954-25G 18954-500MG 18954-5G
76909-532EA 132.62 USD InStock
76909-532 76909-534 76909-536 76909-538
3-Carbamoyl-4-chlorophenylboronic acid ≥97%
3-Carbamoyl-4-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BClNO₃
MW: 199.400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363796
CAS Number: 871332-67-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty