Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-isopropylbenzaldehyde ≥95% research grade

Supplier: AOB CHEM USA
Warning

19423-1G 19423-25G 19423-5G
76909-690EA 84.88 USD InStock
76909-690 76909-692 76909-694
3-Bromo-5-isopropylbenzaldehyde ≥95% research grade
3-Bromo-5-isopropylbenzaldehyde


Formula: C₁₀H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD22543651
CAS Number: 1112210-93-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty