Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylpyridine-2-carboxaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

19814-1G 19814-25G 19814-500MG 19814-5G
76909-824EA 248.26 USD InStock
76909-824 76909-826 76909-828 76909-830
3,5-Dimethylpyridine-2-carboxaldehyde ≥97% research grade
3,5-Dimethylpyridine-2-carboxaldehyde


Formula: C₈H₉NO
MW: 135.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839186
CAS Number: 675138-02-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty