Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-isopropoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

19900-1G 19900-25G 19900-5G
76909-872EA 97.61 USD InStock
76909-872 76909-874 76909-876
3,5-Dichloro-4-isopropoxyphenylboronic acid pinacol ester ≥97%
3,5-Dichloro-4-isopropoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁BCl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD15143609
CAS Number: 1218790-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty