Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,6-dichlorophenol ≥97%

Supplier: AOB CHEM USA
Warning

20160-1G 20160-25G 20160-500MG 20160-5G
76909-968EA 155.96 USD InStock
76909-968 76909-970 76909-972 76909-974
3-Bromo-2,6-dichlorophenol ≥97%
3-Bromo-2,6-dichlorophenol
Precaución: for Research Use Only


Formula: C₆H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD17011993
CAS Number: 116668-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty