Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Difluoro-2-isopropoxyphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20385-1G 20385-25G 20385-5G
76910-128EA 73.21 USD InStock
76910-128 76910-130 76910-132
4,5-Difluoro-2-isopropoxyphenylboronic acid ≥97% research grade
4,5-Difluoro-2-isopropoxyphenylboronic acid


Formula: C₉H₁₁BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09265145
CAS Number: 1072951-61-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty