Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Isopropylbenzeneboronic acid ethylene glycol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

21091-1G 21091-25G 21091-5G
76910-344EA 62.6 USD InStock
76910-344 76910-346 76910-348
3-Isopropylbenzeneboronic acid ethylene glycol ester ≥97% research grade
3-Isopropylbenzeneboronic acid ethylene glycol ester


Formula: C₁₁H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD02093946
CAS Number: 374537-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty