Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dibromo-1H-indazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

22164-1G 22164-25G 22164-500MG 22164-5G
76910-738EA 328.88 USD InStock
76910-738 76910-740 76910-742 76910-744
4,7-Dibromo-1H-indazole ≥97% research grade
4,7-Dibromo-1H-indazole


Formula: C₇H₄Br₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD19687064
CAS Number: 1316273-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty