Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-pyrrolidinophenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

22174-1G 22174-25G 22174-500MG 22174-5G
76910-750EA 153.84 USD InStock
76910-750 76910-752 76910-754 76910-756
3-Fluoro-5-pyrrolidinophenylboronic acid ≥97% research grade
3-Fluoro-5-pyrrolidinophenylboronic acid


Formula: C₁₀H₁₃BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD12546542
CAS Number: 1217500-96-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty