Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-6-ethoxy-2-fluorophenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

22272-1G 22272-25G 22272-500MG 22272-5G
76910-766EA 144.29 USD InStock
76910-766 76910-768 76910-770 76910-772
(3-Bromo-6-ethoxy-2-fluorophenyl)boronic acid ≥97% research grade
(3-Bromo-6-ethoxy-2-fluorophenyl)boronic acid


Formula: C₈H₉BBrFO₃
MW: 262.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026724
CAS Number: 1309980-98-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty