Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

22356-1G 22356-25G 22356-500MG 22356-5G
76910-780EA 201.58 USD InStock
76910-780 76910-782 76910-784 76910-786
3,5-Difluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥97% research grade
3,5-Difluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid


Formula: C₁₁H₁₂BF₂NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20526377
CAS Number: 1264616-16-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty