Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-methoxycarbonylbenzeneboronic acid, pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

22638-1G 22638-25G 22638-5G
76910-900EA 88.06 USD InStock
76910-900 76910-902 76910-904
3-Fluoro-5-methoxycarbonylbenzeneboronic acid, pinacol ester ≥97% research grade
3-Fluoro-5-methoxycarbonylbenzeneboronic acid, pinacol ester


Formula: C₁₄H₁₈BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD12546616
CAS Number: 1016979-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty