Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloropyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

22769-1G 22769-500MG 22769-25G 22769-5G
76910-984EA 201.58 USD InStock
76910-984 76910-986 76927-868 76927-870
4-Bromo-2-chloropyridine-3-boronic acid pinacol ester ≥97%
4-Bromo-2-chloropyridine-3-boronic acid pinacol ester


Formula: C₁₁H₁₄BBrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16995521
CAS Number: 2121514-27-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty