Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Butoxy-5-formylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

22898-1G 22898-25G 22898-500MG 22898-5G
76911-064EA 231.28 USD InStock
76911-064 76911-066 76911-068 76911-070
3-Butoxy-5-formylphenylboronic acid ≥97%
3-Butoxy-5-formylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅BO₄
Storage Temperature: Ambient
MDL Number: MFCD08705271
CAS Number: 1072951-70-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty