Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyano-5-(trifluoromethyl)benzoic Acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

23502-1G 23502-25G 23502-5G
76911-288EA 97.61 USD InStock
76911-288 76911-290 76911-292
3-Cyano-5-(trifluoromethyl)benzoic Acid ≥95% research grade
3-Cyano-5-(trifluoromethyl)benzoic Acid


Formula: C₉H₄F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD16610738
CAS Number: 942077-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty