Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2,4-Difluorophenyl)benzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

23926-1G 23926-25G 23926-500MG 23926-5G
76911-426EA 231.28 USD InStock
76911-426 76911-428 76911-430 76911-432
4-(2,4-Difluorophenyl)benzaldehyde ≥97% research grade
4-(2,4-Difluorophenyl)benzaldehyde


Formula: C₁₃H₈F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD04117388
CAS Number: 728918-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty