Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminopyridine-3-boronic acid, pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

24350-25G 24350-5G
76911-552EA 445.58 USD InStock
76911-552 76911-554
5-Aminopyridine-3-boronic acid, pinacol ester ≥97%
5-Aminopyridine-3-boronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₇BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09260489
CAS Number: 1073354-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty