Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-ethylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

24606-1G 24606-25G 24606-500MG 24606-5G
76911-718EA 185.66 USD InStock
76911-718 76911-720 76911-722 76911-724
3-Bromo-5-ethylbenzoic acid ≥97% research grade
3-Bromo-5-ethylbenzoic acid


Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01463856
CAS Number: 342045-08-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty