Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopropyl-2,6-difluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

25643-1G 25643-25G 25643-500MG 25643-5G
76912-020EA 439.22 USD InStock
76912-020 76912-022 76912-024 76912-026
3-Cyclopropyl-2,6-difluorophenylboronic acid ≥97%
3-Cyclopropyl-2,6-difluorophenylboronic acid


Formula: C₉H₉BF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD23713139
CAS Number: 1451393-02-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty