Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥97% research grade

Supplier: AOB CHEM USA
Warning

25875-1G 25875-25G 25875-500MG 25875-5G
76912-096EA 290.69 USD InStock
76912-096 76912-098 76912-100 76912-102
4-(Trifluoromethoxy)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥97% research grade
4-(Trifluoromethoxy)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline


Formula: C₁₃H₁₇BF₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD16996387
CAS Number: 1451391-22-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty