Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-4-(hydroxymethyl)phenyl)boronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

26292-1G 26292-25G 26292-500MG 26292-5G
76912-300EA 230.22 USD InStock
76912-300 76912-302 76912-304 76912-306
(3-Chloro-4-(hydroxymethyl)phenyl)boronic acid ≥95% research grade
(3-Chloro-4-(hydroxymethyl)phenyl)boronic acid


Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18384123
CAS Number: 1190875-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty