Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

26322-1G 26322-25G 26322-500MG 26322-5G
76912-310EA 265.23 USD InStock
76912-310 76912-312 76912-314 76912-316
3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥97% research grade
3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester


Formula: C₁₄H₁₉BO₅
MW: 278.108 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546597
CAS Number: 1004294-79-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty